Assay Details
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243883.1 | NP_001230812.1 | 1 - 2 | 541 | 62 | |
GenBank mRNA | AJ236934.1 | - | 1 - 2 | 501 | 62 | |
AK231339.1 | - | 1 - 2 | 541 | 62 | ||
AK237000.1 | - | 1 - 2 | 507 | 62 | ||
AK344422.1 | - | 1 - 2 | 340 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
