Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243887.1 | NP_001230816.1 | 1 - 2 | 109 | 59 | |
| GenBank mRNA | AK239819.1 | - | 1 - 2 | 109 | 59 | |
| AK395834.1 | - | 1 - 2 | 121 | 59 | ||
| AK400888.1 | - | 1 - 2 | 109 | 59 | ||
More Information
Set Membership: |
|

FAM32A
Genomic Map