Assay Details
| Gene Symbol: SET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244090.1 | NP_001231019.1 | 5 - 6 | 693 | 84 | |
| XM_021071997.1 | XP_020927656.1 | 643 | 84 | |||
| XM_021072036.1 | XP_020927695.1 | 641 | 84 | |||
| XM_021072067.1 | XP_020927726.1 | 844 | 84 | |||
| GenBank mRNA | AK347372.1 | - | 5 - 6 | 693 | 84 | |
| AK351041.1 | - | 5 - 6 | 604 | 84 | ||
More Information
Set Membership: |
|

SET
Genomic Map