Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244208.1 | NP_001231137.1 | 3 - 4 | 641 | 82 | |
| GenBank mRNA | AK238807.1 | - | 3 - 4 | 639 | 82 | |
| AK351591.1 | - | 3 - 4 | 606 | 82 | ||
| AK351936.1 | - | 3 - 4 | 599 | 82 | ||
| AK392733.1 | - | 3 - 4 | 614 | 82 | ||
| AK396232.1 | - | 3 - 4 | 599 | 82 | ||
| AK397156.1 | - | 3 - 4 | 604 | 82 | ||
| AK397840.1 | - | 3 - 4 | 641 | 82 | ||
| AK398185.1 | - | 3 - 4 | 599 | 82 | ||
| CJ022605.1 | - | 3 - 4 | 641 | 82 | ||
| DB817925.1 | - | 2 - 3 | 630 | 82 | ||
More Information
Set Membership: |
|

PRELID1
Genomic Map