Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244220.1 | NP_001231149.1 | 2 - 3 | 497 | 65 | |
| GenBank mRNA | AK346041.1 | - | 2 - 3 | 480 | 65 | |
| AK346392.1 | - | 2 - 3 | 497 | 65 | ||
| BX915266.2 | - | 1 - 2 | 167 | 65 | ||
More Information
Set Membership: |
|

ZCCHC9
Genomic Map