Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244336.1 | NP_001231265.1 | 2 - 3 | 454 | 83 | |
| GenBank mRNA | AK237473.1 | - | 2 - 3 | 454 | 83 | |
| AK239833.1 | - | 1 - 2 | 525 | 83 | ||
| AK394359.1 | - | 2 - 3 | 458 | 83 | ||
| AY610118.1 | - | 2 - 3 | 454 | 83 | ||
More Information
Set Membership: |
|

TMUB1
Genomic Map