Assay Details
| Gene Symbol: MRPL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244380.1 | NP_001231309.1 | 2 - 3 | 278 | 60 | |
| GenBank mRNA | AK399780.1 | - | 2 - 3 | 272 | 60 | |
| AY609942.1 | - | 2 - 3 | 278 | 60 | ||
| BQ598943.1 | - | 1 - 2 | 554 | 60 | ||
More Information
Set Membership: |
|

MRPL17
Genomic Map