Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244505.1 | NP_001231434.1 | 1 - 2 | 175 | 83 | |
| GenBank mRNA | AK232094.1 | - | 1 - 2 | 146 | 83 | |
| AK395975.1 | - | 1 - 2 | 149 | 83 | ||
| DT324394.1 | - | 1 - 2 | 175 | 83 | ||
More Information
Set Membership: |
|

TNFAIP8
Genomic Map