Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244623.1 | NP_001231552.1 | 10 - 11 | 1794 | 72 | |
| XM_005655347.2 | XP_005655404.1 | 72 | ||||
| XM_005655348.2 | XP_005655405.1 | 72 | ||||
| GenBank mRNA | AK236645.1 | - | 10 - 11 | 1370 | 72 | |
| AK348996.1 | - | 6 - 7 | 72 | |||
| EW479761.2 | - | 2 - 3 | 72 | |||
More Information
Set Membership: |
|

MTDH
Genomic Map