Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244711.1 | NP_001231640.1 | 3 - 4 | 236 | 75 | |
| XM_021066097.1 | XP_020921756.1 | 372 | 75 | |||
| XM_021066098.1 | XP_020921757.1 | 163 | 75 | |||
| XM_021066099.1 | XP_020921758.1 | 265 | 75 | |||
| GenBank mRNA | AK239277.1 | - | 3 - 4 | 236 | 75 | |
| AK398433.1 | - | 857 | 75 | |||
| AY610081.1 | - | 3 - 4 | 178 | 75 | ||
More Information
Set Membership: |
|

LIMD2
Genomic Map