Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244730.1 | NP_001231659.1 | 4 - 5 | 535 | 77 | |
| XM_005653697.3 | XP_005653754.1 | 600 | 77 | |||
| XM_005653698.3 | XP_005653755.1 | 2762 | 77 | |||
| XM_005653699.3 | XP_005653756.1 | 591 | 77 | |||
| XM_005653701.3 | XP_005653758.1 | 630 | 77 | |||
| XM_013977422.2 | XP_013832876.1 | 1074 | 77 | |||
| XM_013977439.2 | XP_013832893.1 | 670 | 77 | |||
| XM_013977441.1 | XP_013832895.1 | 680 | 77 | |||
| XM_021102035.1 | XP_020957694.1 | 1137 | 77 | |||
| XM_021102036.1 | XP_020957695.1 | 1332 | 77 | |||
| GenBank mRNA | AK238962.1 | - | 4 - 5 | 535 | 77 | |
| AK389867.1 | - | 4 - 5 | 506 | 77 | ||
| AK398411.1 | - | 2 - 3 | 913 | 77 | ||
More Information
Set Membership: |
|

JAML
Genomic Map