Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244730.1 | NP_001231659.1 | 8 - 9 | 1078 | 73 | |
| XM_005653697.3 | XP_005653754.1 | 1143 | 73 | |||
| XM_005653698.3 | XP_005653755.1 | 3305 | 73 | |||
| XM_005653699.3 | XP_005653756.1 | 1134 | 73 | |||
| XM_005653701.3 | XP_005653758.1 | 1173 | 73 | |||
| XM_013977422.2 | XP_013832876.1 | 1617 | 73 | |||
| XM_013977439.2 | XP_013832893.1 | 1213 | 73 | |||
| XM_013977441.1 | XP_013832895.1 | 1223 | 73 | |||
| XM_021102035.1 | XP_020957694.1 | 1680 | 73 | |||
| XM_021102036.1 | XP_020957695.1 | 1875 | 73 | |||
| GenBank mRNA | AK238962.1 | - | 8 - 9 | 1078 | 73 | |
| AK398411.1 | - | 6 - 7 | 1456 | 73 | ||
More Information
Set Membership: |
|

JAML
Genomic Map