Assay Details
Gene Symbol: MIR182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128422.1 | 1 | 44 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-96 | MIR96 | ssc-mir-96 | -4.2kb | Sense | Chr. 18 - 18982506 - 18982590 on NCBI Build Sscrofa11.1 |
Downstream | nuclear respiratory factor 1 | NRF1 | 21.5kb | Anti-sense | Chr. 18 - 19008408 - 19142678 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
