Assay Details
| Gene Symbol: MIRLET7E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038521.1 | 1 | 40 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-125a | MIR125A | ssc-mir-125a | -397.0 | Sense | Chr. 6 - 58331566 - 58331645 on NCBI Build Sscrofa11.1 |
| Downstream | microRNA mir-99b | MIR99B | ssc-mir-99b | 90.0 | Sense | Chr. 6 - 58332211 - 58332280 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIRLET7E
Genomic Map