Assay Details
Gene Symbol: MIR9813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128498.1 | 1 | 33 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium voltage-gated channel subfamily A member 5 | KCNA5 | -37.4kb | Anti-sense | Chr. 5 - 65439336 - 65442207 on NCBI Build Sscrofa11.1 | |
Downstream | potassium voltage-gated channel subfamily A member 1 | KCNA1 | 77.2kb | Anti-sense | Chr. 5 - 65556837 - 65565880 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
