Assay Details
Gene Symbol: MIR9824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128509.1 | 1 | 38 | 49 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
afadin, adherens junction formation factor |
AFDN | Sense | Chr. 1 - 1569652 - 1698475 on NCBI Build Sscrofa11.1 | ||
Downstream | uncharacterized LOC106506286 | LOC106506286 | 151.0kb | Anti-sense | Chr. 1 - 1816683 - 1835173 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
