Assay Details
Gene Symbol: MIR490-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038585.1 | 1 | 38 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pleiotrophin | PTN | PTF-BETA | -259.2kb | Anti-sense | Chr. 18 - 12219708 - 12309045 on NCBI Build Sscrofa11.1 |
Downstream | myotrophin | MTPN | 761.8kb | Anti-sense | Chr. 18 - 13330049 - 13395297 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
