Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244688.1 | NP_001231617.1 | 8 - 9 | 1138 | 94 | |
| XM_021093303.1 | XP_020948962.1 | 1069 | 94 | |||
| GenBank mRNA | AK238066.1 | - | 8 - 9 | 1138 | 94 | |
More Information
Set Membership: |
|

CCDC113
Genomic Map