Assay Details
| Gene Symbol: FBXO34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258439.1 | NP_001245368.1 | 1 | 2715 | 109 | |
| XM_021067261.1 | XP_020922920.1 | 2948 | 109 | |||
| XM_021067308.1 | XP_020922967.1 | 2885 | 109 | |||
| XM_021067382.1 | XP_020923041.1 | 3166 | 109 | |||
| XM_021067448.1 | XP_020923107.1 | 4060 | 109 | |||
| XM_021067475.1 | XP_020923134.1 | 3101 | 109 | |||
| XM_021067524.1 | XP_020923183.1 | 3038 | 109 | |||
| XM_021067582.1 | XP_020923241.1 | 3028 | 109 | |||
| XM_021067659.1 | XP_020923318.1 | 2952 | 109 | |||
| XM_021067706.1 | XP_020923365.1 | 3923 | 109 | |||
| GenBank mRNA | AK391616.1 | - | 1 | 2423 | 109 | |
More Information
Set Membership: |
|

FBXO34
Genomic Map