Assay Details
| Gene Symbol: NKX2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359915.4 | XP_003359963.1 | 1524 | 80 | ||
| XM_021077994.1 | XP_020933653.1 | 1527 | 80 | |||
More Information
Set Membership: |
|

NKX2-2
Genomic Map