Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481711.4 | XP_003481759.2 | 1354 | 64 | ||
| XM_003481712.4 | XP_003481760.2 | 7252 | 64 | |||
More Information
Set Membership: |
|

PDZRN4
Genomic Map