Assay Details
| Gene Symbol: TAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005654524.3 | XP_005654581.2 | 322 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005654524.3 transcripts.
Set Membership: |
|

TAL2
Genomic Map