Assay Details
| Gene Symbol: PGPEP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659773.3 | XP_005659830.1 | 752 | 64 | ||
| XM_005659775.3 | XP_005659832.1 | 755 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGPEP1L| Gene Symbol: PGPEP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659773.3 | XP_005659830.1 | 752 | 64 | ||
| XM_005659775.3 | XP_005659832.1 | 755 | 64 | |||
Set Membership: |
|