Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661521.3 | XP_005661578.1 | 873 | 122 | ||
| XM_005661522.3 | XP_005661579.1 | 451 | 122 | |||
| XM_005661523.3 | XP_005661580.1 | 574 | 122 | |||
| XM_021084497.1 | XP_020940156.1 | 448 | 122 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005661521.3, XM_005661522.3, XM_005661523.3, XM_021084497.1 transcripts.
Set Membership: |
|

ANKRD34B
Genomic Map