Assay Details
| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664855.3 | XP_005664912.1 | 2676 | 65 | ||
| XM_005664858.3 | XP_005664915.1 | 2505 | 65 | |||
| XM_013998807.2 | XP_013854261.1 | 3017 | 65 | |||
More Information
Set Membership: |
|

NLRP9
Genomic Map