Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669658.3 | XP_005669715.1 | 641 | 90 | ||
| XM_005669660.3 | XP_005669717.1 | 506 | 90 | |||
| XM_013981798.2 | XP_013837252.1 | 1856 | 90 | |||
More Information
Set Membership: |
|

WNT5A
Genomic Map