Assay Details
| Gene Symbol: ATOH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005671039.3 | XP_005671096.1 | 1021 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005671039.3 transcripts.
Set Membership: |
|

ATOH7
Genomic Map