Assay Details
| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995792.2 | XP_013851246.2 | 393 | 71 | ||
| XM_013995793.2 | XP_013851247.2 | 393 | 71 | |||
| XM_021086996.1 | XP_020942655.1 | 393 | 71 | |||
| XM_021086997.1 | XP_020942656.1 | 393 | 71 | |||
More Information
Set Membership: |
|

BARX1
Genomic Map