Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063724.1 | XP_020919383.1 | 1188 | 67 | ||
| XM_021063725.1 | XP_020919384.1 | 521 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063724.1 | XP_020919383.1 | 1188 | 67 | ||
| XM_021063725.1 | XP_020919384.1 | 521 | 67 | |||
Set Membership: |
|