Assay Details
| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070067.1 | XP_020925726.1 | 76 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021070067.1 transcripts.
Set Membership: |
|

LRRC15
Genomic Map