Assay Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093029.1 | XP_020948688.1 | 1040 | 78 | ||
| XM_021093030.1 | XP_020948689.1 | 4229 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093029.1 | XP_020948688.1 | 1040 | 78 | ||
| XM_021093030.1 | XP_020948689.1 | 4229 | 78 | |||
Set Membership: |
|