Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132779.5 | XP_003132827.4 | 1153 | 66 | ||
| XM_021071007.1 | XP_020926666.1 | 997 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132779.5 | XP_003132827.4 | 1153 | 66 | ||
| XM_021071007.1 | XP_020926666.1 | 997 | 66 | |||
Set Membership: |
|