Assay Details
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134165.4 | XP_003134213.1 | 751 | 53 | ||
| XM_021076892.1 | XP_020932551.1 | 843 | 53 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134165.4 | XP_003134213.1 | 751 | 53 | ||
| XM_021076892.1 | XP_020932551.1 | 843 | 53 | |||
Set Membership: |
|