Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353290.4 | XP_003353338.1 | 289 | 112 | ||
| XM_005659391.2 | XP_005659448.1 | 285 | 112 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003353290.4, XM_005659391.2 transcripts.
Set Membership: |
|

FAM229B
Genomic Map