Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353290.4 | XP_003353338.1 | 426 | 101 | ||
| XM_003353291.3 | XP_003353339.1 | 265 | 101 | |||
| XM_005659391.2 | XP_005659448.1 | 422 | 101 | |||
| XM_005659393.2 | XP_005659450.1 | 343 | 101 | |||
| XM_021091408.1 | XP_020947067.1 | 367 | 101 | |||
More Information
Set Membership: |
|

FAM229B
Genomic Map