Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998435.2 | XP_013853889.1 | 413 | 102 | ||
| XM_013998436.2 | XP_013853890.1 | 535 | 102 | |||
| XM_013998437.2 | XP_013853891.1 | 567 | 102 | |||
| XM_013998438.2 | XP_013853892.1 | 610 | 102 | |||
| XM_013998439.2 | XP_013853893.1 | 1637 | 102 | |||
| XM_021097233.1 | XP_020952892.1 | 528 | 102 | |||
| XM_021097235.1 | XP_020952894.1 | 488 | 102 | |||
More Information
Set Membership: |
|

ZNF527
Genomic Map