Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669819.3 | XP_005669876.1 | 1944 | 85 | ||
| XM_013982012.2 | XP_013837466.1 | 1944 | 85 | |||
More Information
Set Membership: |
|

KBTBD12
Genomic Map