Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123056.5 | XP_003123104.1 | 165 | 69 | ||
| GenBank mRNA | AK233221.1 | - | 1 - 2 | 151 | 69 | |
| AK395605.1 | - | 1 - 2 | 154 | 69 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map