Assay Details
| Gene Symbol: AMDHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092739.1 | XP_020948398.1 | 610 | 84 | ||
| GenBank mRNA | AK233515.1 | - | 4 - 5 | 643 | 84 | |
| AK394151.1 | - | 4 - 5 | 642 | 84 | ||
More Information
Set Membership: |
|

AMDHD1
Genomic Map