Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092596.1 | XP_020948255.1 | 1082 | 57 | ||
| XM_021092606.1 | XP_020948265.1 | 1011 | 57 | |||
| XM_021092609.1 | XP_020948268.1 | 1040 | 57 | |||
| XM_021092612.1 | XP_020948271.1 | 953 | 57 | |||
More Information
Set Membership: |
|

ST8SIA5
Genomic Map