Assay Details
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663044.3 | XP_005663101.1 | 701 | 74 | ||
| XM_005663046.3 | XP_005663103.1 | 641 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663044.3 | XP_005663101.1 | 701 | 74 | ||
| XM_005663046.3 | XP_005663103.1 | 641 | 74 | |||
Set Membership: |
|