Assay Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094797.1 | XP_020950456.1 | 280 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021094797.1 transcripts.
Set Membership: |
|

LRRC4B
Genomic Map