Assay Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978966.2 | XP_013834420.2 | 1581 | 93 | ||
| XM_013978981.2 | XP_013834435.2 | 1456 | 93 | |||
| XM_013978986.2 | XP_013834440.2 | 1188 | 93 | |||
| XM_013978992.2 | XP_013834446.2 | 727 | 93 | |||
| XM_021065976.1 | XP_020921635.1 | 729 | 93 | |||
| XM_021065977.1 | XP_020921636.1 | 1308 | 93 | |||
| XM_021065978.1 | XP_020921637.1 | 973 | 93 | |||
| XM_021065980.1 | XP_020921639.1 | 1122 | 93 | |||
| XM_021065981.1 | XP_020921640.1 | 1575 | 93 | |||
| XM_021065982.1 | XP_020921641.1 | 1575 | 93 | |||
More Information
Set Membership: |
|

DCUN1D2
Genomic Map