Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091439.1 | XP_020947098.1 | 478 | 85 | ||
| XM_021091440.1 | XP_020947099.1 | 489 | 85 | |||
More Information
Set Membership: |
|

TNFRSF13C
Genomic Map