Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095116.1 | XP_020950775.1 | 417 | 92 | ||
| XM_021095117.1 | XP_020950776.1 | 502 | 92 | |||
| XM_021095118.1 | XP_020950777.1 | 447 | 92 | |||
| XM_021095119.1 | XP_020950778.1 | 608 | 92 | |||
| XM_021095120.1 | XP_020950779.1 | 1042 | 92 | |||
More Information
Set Membership: |
|

ZNF444
Genomic Map