Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121398.6 | XP_003121446.3 | 1059 | 104 | ||
| XM_021092140.1 | XP_020947799.1 | 576 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121398.6 | XP_003121446.3 | 1059 | 104 | ||
| XM_021092140.1 | XP_020947799.1 | 576 | 104 | |||
Set Membership: |
|