Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083871.1 | XP_020939530.1 | 1342 | 65 | ||
| XM_021083872.1 | XP_020939531.1 | 1332 | 65 | |||
| GenBank mRNA | KF017598.1 | - | 1 - 2 | 299 | 65 | |
More Information
Set Membership: |
|

ANGPTL8
Genomic Map