Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083872.1 | XP_020939531.1 | 1606 | 54 | ||
| GenBank mRNA | KF017598.1 | - | 3 - 4 | 573 | 54 | |
| KX447621.1 | - | 3 - 4 | 591 | 54 | ||
More Information
Set Membership: |
|

ANGPTL8
Genomic Map