Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977531.2 | XP_013832985.2 | 63 | 121 | ||
| GenBank mRNA | AK233896.1 | - | 1 - 2 | 32 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977531.2 transcripts.
Set Membership: |
|

ZSCAN16
Genomic Map