Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665669.3 | XP_005665726.2 | 1247 | 83 | ||
| XM_005665670.3 | XP_005665727.2 | 1165 | 83 | |||
| XM_013977531.2 | XP_013832985.2 | 661 | 83 | |||
| GenBank mRNA | AK233896.1 | - | 3 - 4 | 630 | 83 | |
More Information
Set Membership: |
|

ZSCAN16
Genomic Map